CAS RN 2320274-78-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(RS)-Butyryltimolol, MedChemExpress
MedChemExpress (RS)-Butyryltimolol is the racemate of Butyryltimolol. Butyryltimolol, an effective prodrug of Timolol, improves the corneal penetration of Timolol. Butyryltimolol is a β-adrenergic blocker.