CAS RN 2310085-85-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ BIBO 3304 trifluoroacetate
Highly selective NPY Y1 receptor antagonist