CAS RN 230963-27-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4-O-Acetyl-3,6-di-O-(3,4,6-tri-O-acetyl-Beta-D-mannopyranosyl-ethylidyne)-1,2-O-ethylidene-Beta-D-mannopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-O-Acetyl-3,6-di-O-(3,4,6-tri-O-acetyl-Beta-D-mannopyranosyl-ethylidyne)-1,2-O-ethylidene-Beta-D-mannopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-O-Acetyl-3,6-di-O-(3,4,6-tri-O-acetyl-Beta-D-mannopyranosyl-ethylidyne)-1,2-O-ethylidene-Beta-D-mannopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.