CAS RN 2304-98-5
IUPAC Navn:
5-{[amino(nitroamino)methylidene]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonymer:
5-{[amino(nitroamino)methylidene]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molekylvægt (g/mol):
353.34
Molekylformel:
C14H19N5O6
InChi Key:
BZPCSFNCKORLQG-UHFFFAOYNA-N
SMILES:
NC(N[N+]([O-])=O)=NCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
af
1
Resultater
1
Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine, 98%, Thermo Scientific™
CAS: 2304-98-5 Molekylær formel: C14H19N5O6 Molekylvægt (g/mol): 353.335 MDL nummer: MFCD00038092 InChI nøgle: BZPCSFNCKORLQG-NSHDSACASA-N Synonym: z-arg no2-oh,z-l-arg no2-oh,n-cbz-n-nitro-l-arginine,cbz-l-arg no2-oh,s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid,l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl,cbz-arg no2-oh,n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine,2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid,n-alpha-z-n-omega-nitro-l-arginine PubChem CID: 12695023 SMIL: C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O