CAS RN 2250025-93-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ Clozapine N-oxide dihydrochloride
Activator of muscarinic DREADDs; water soluble version of Clozapine N-oxide (Cat. No. 4936)