CAS RN 2237942-08-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
MC180295, MedChemExpress
MedChemExpress MC180295 ((rel)-MC180295) is a potent and selective CDK9-Cyclin T1 inhibitor, with an IC50 of 5 nM, at least 22-fold more selective for CDK9 over other CDKs. MC180295 also inhibits GSK-3α and GSK-3β. MC180295 ((rel)-MC180295) has potent anti-tumor effect.