CAS RN 223387-33-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(2R,2'R,3R,3'R,4R)-2,2'-Bis[3,4-bis(phenylmethoxy)phenyl]-3,3',4,4'-tetrahydro-5,5',7,7'-tetrakis(phenylmethoxy)[4,8'-bi-2H-1-benzopyran]-3,3'-diyl Ester 3,4,5-Tris(phenylmethoxy)-benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R,2'R,3R,3'R,4R)-2,2'-Bis[3,4-bis(phenylmethoxy)phenyl]-3,3',4,4'-tetrahydro-5,5',7,7'-tetrakis(phenylmethoxy)[4,8'-bi-2H-1-benzopyran]-3,3'-diyl Ester 3,4,5-Tris(phenylmethoxy)-benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2R,2'R,3R,3'R,4R)-2,2'-Bis[3,4-bis(phenylmethoxy)phenyl]-3,3',4,4'-tetrahydro-5,5',7,7'-tetrakis(phenylmethoxy)[4,8'-bi-2H-1-benzopyran]-3,3'-diyl Ester 3,4,5-Tris(phenylmethoxy)-benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.