CAS RN 22316-55-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
N-Desmethyl Clobazam, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Chloro-5-phenyl-1,5-dihydro-3H-1,5-benzodiazepine-2,4-dione (Norclobazam), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
N-Desmethyl Clobazam, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.