CAS RN 2225940-48-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ Pomalidomide 4'-alkylC4-acid
Cereblon ligand with alkyl linker and terminal acid for onward chemistry