CAS RN 2213-23-2
IUPAC Navn:
2,4-dimethylheptane
Synonymer:
heptane, 2,4-dimethyl-
Molekylvægt (g/mol):
128.26
Molekylformel:
C9H20
InChi Key:
AUKVIBNBLXQNIZ-UHFFFAOYNA-N
SMILES:
CCCC(C)CC(C)C
1
–
1
af
1
Resultater
1
ASTM Method D5134 Linearity Check Mixture, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality