CAS RN 220119-16-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
25-Cyclohexyl-5-demethoxy-25-de(1-methylpropyl)-22,23-dihydro-5-oxo-avermectin A1a, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
25-Cyclohexyl-5-demethoxy-25-de(1-methylpropyl)-22,23-dihydro-5-oxo-avermectin A1a, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
25-Cyclohexyl-5-demethoxy-25-de(1-methylpropyl)-22,23-dihydro-5-oxo-avermectin A1a, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.