CAS RN 2172820-09-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ VH 032 amide-PEG2-acid
VHL ligand with PEG linker and terminal acid for onward chemistry