CAS RN 2170106-13-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
4,4'-(1H-1,2,4-Triazol-1-ylmethylene)dibenzamide, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Letrozole di-Amide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Letrozole di-Amide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Letrozole di-Amide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.