CAS RN 2169266-70-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ Thalidomide 4'-ether-alkylC7-acid
Cereblon ligand with alkyl linker and terminal acid for onward chemistry