CAS RN 216167-92-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Camobucol, MedChemExpress
MedChemExpress Camobucol (AGIX 4207) is an orally active, phenolic antioxidant and anti-inflammatory compound with antirheumatic properties.