CAS RN 2156588-26-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ ARCC 4 negative control
Negative control for ARCC 4 (Cat. No. 7254)