CAS RN 212332-35-9
IUPAC Navn:
[4-(5-ethoxypyridin-3-yl)but-3-en-1-yl](methyl)amine; bis(but-2-enedioic acid)
Synonymer:
[4-(5-ethoxypyridin-3-yl)but-3-en-1-yl](methyl)amine; bis(butenedioic acid)
Molekylvægt (g/mol):
438.43
Molekylformel:
C20H26N2O9
InChi Key:
GEWVPSJQGJBDLM-UHFFFAOYSA-N
SMILES:
OC(=O)C=CC(O)=O.OC(=O)C=CC(O)=O.CCOC1=CN=CC(C=CCCNC)=C1
1
–
4
af
4
Resultater
1
TC 2559 Fumarate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
TC 2559 Fumarate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
TC 2559 Fumarate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Tocris Bioscience™ TC 2559 difumarate
CAS: 212332-35-9 Molekylær formel: C20H26N2O9