CAS RN 2116542-23-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
7-[4-(4-Benzo[b]thien-4-yl-1-piperazinyl)butoxy]-1-[4-[(1,2,3,4-tetrahydro-2-oxo-7-quinolinyl)oxy]butyl]-2(1H)-quinolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-[4-(4-Benzo[b]thien-4-yl-1-piperazinyl)butoxy]-1-[4-[(1,2,3,4-tetrahydro-2-oxo-7-quinolinyl)oxy]butyl]-2(1H)-quinolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-[4-(4-Benzo[b]thien-4-yl-1-piperazinyl)butoxy]-1-[4-[(1,2,3,4-tetrahydro-2-oxo-7-quinolinyl)oxy]butyl]-2(1H)-quinolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.