CAS RN 2095832-33-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
MedChemExpress DMA trihydrochloride
MedChemExpress DMA trihydrochloride is a fluorescent compound (λex=340 nm, λem=478 nm).