CAS RN 2095485-22-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ BTD S-sulfenic acid probe
Chemoproteomic reagent for targeting S-sulfenic acids on SulfenQ platform, alkyne handle for click chemistry