CAS RN 2095417-18-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
3'-Azido-3'-deoxy-5-fluorocytidine, MedChemExpress
MedChemExpress 3'-Azido-3'-deoxy-5-fluorocytidine (Compound 12) is a cytidine derivative.