CAS RN 2093120-32-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ PA Janelia Fluor 646, SE
Red photoactivatable dye (Reactive group: NHS ester). Application: super resolution microscopy (SRM), single molecule tracking and live cell imaging