CAS RN 2053424-82-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
D-Alanine N-[(S)-[[(1R)-2-(6-Aamino-9Hpurin-9-yl)-1-methylethoxy]methyl]phenoxyphosphinyl]-1-methylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
D-Alanine N-[(S)-[[(1R)-2-(6-Aamino-9Hpurin-9-yl)-1-methylethoxy]methyl]phenoxyphosphinyl]-1-methylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
D-Alanine N-[(S)-[[(1R)-2-(6-Aamino-9Hpurin-9-yl)-1-methylethoxy]methyl]phenoxyphosphinyl]-1-methylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.