CAS RN 204204-75-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(3E)-4-(Acetylthio)-Alpha-(2-chlorophenyl)-3-(2-ethoxy-2-oxoethylidene)-1-piperidineacetic Acid Methyl Ester(Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3E)-4-(Acetylthio)-Alpha-(2-chlorophenyl)-3-(2-ethoxy-2-oxoethylidene)-1-piperidineacetic Acid Methyl Ester(Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.