CAS RN 2026-58-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Alpha-Phenyl-Alpha-[3-(phenyl-2-pyridinylmethylene)-2,4-cyclopentadien-1-yl]-2-pyridinemethanol (E/Z mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
alpha-Phenyl-alpha-[3-(phenyl-2-pyridinylmethylene)-2,4-cyclopentadien-1-yl]-2-pyridinemethanol (E/Z mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Alpha-Phenyl-Alpha-[3-(phenyl-2-pyridinylmethylene)-2,4-cyclopentadien-1-yl]-2-pyridinemethanol (E/Z mixture), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.