CAS RN 201870-82-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(Acetato-KappaO)[[2,2'-[(1R,2R)-1,2-cyclohexanediylbis[(nitrilo-KappaN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-KappaO]](2-)]cobalt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(Acetato-KappaO)[[2,2'-[(1R,2R)-1,2-cyclohexanediylbis[(nitrilo-KappaN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-KappaO]](2-)]cobalt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.