CAS RN 2012523-61-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
3-(4-Phenoxyphenyl)-1-[(3R)-1-(phenylmethyl)-3-piperidinyl]-1H-pyrazolo[3,4-d]pyrimidin-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(4-Phenoxyphenyl)-1-[(3R)-1-(phenylmethyl)-3-piperidinyl]-1H-pyrazolo[3,4-d]pyrimidin-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-(4-Phenoxyphenyl)-1-[(3R)-1-(phenylmethyl)-3-piperidinyl]-1H-pyrazolo[3,4-d]pyrimidin-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.