CAS RN 2011720-21-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ PA Janelia Fluor™ 646, Haloalkane
Photoactivatable Janelia Fluor™ 646 with chloroalkane handle for self-labeling tags fusion protein