CAS RN 1944-63-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
1,5-Dioxo-7aBeta-methyl-3aAlpha-hexahydroindane-4Alpha-propionic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,5-Dioxo-7aBeta-methyl-3aAlpha-hexahydroindane-4Alpha-propionic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,5-Dioxo-7aBeta-methyl-3aAlpha-hexahydroindane-4Alpha-propionic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.