CAS RN 1923194-81-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
11,11′-[Ethylenebis(oxyethylenepiperazine-4,1-diyl)]bis(dibenzo[b,f][1,4]thiazepine) Tetrahydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.