CAS RN 189035-07-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(11Beta,17Beta)-11-(1,3-Benzodioxol-5-yl)-17-hydroxy-17-(1-propyn-1-yl)estra-4,9-dien-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(11b,17b)-11-(1,3-Benzodioxol-5-yl)-17-hydroxy-17-(1-propyn-1-yl)estra-4,9-dien-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(11Beta,17Beta)-11-(1,3-Benzodioxol-5-yl)-17-hydroxy-17-(1-propyn-1-yl)estra-4,9-dien-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.