CAS RN 18625-12-2
IUPAC Navn:
methyl 4-(2,4-dichlorophenoxy)butanoate
Synonymer:
methyl 4-(2,4-dichlorophenoxy)butanoate
Molekylvægt (g/mol):
263.11
Molekylformel:
C11H12Cl2O3
InChi Key:
NKXSNMCGZZWMMW-UHFFFAOYSA-N
SMILES:
COC(=O)CCCOC1=CC=C(Cl)C=C1Cl
1
–
1
af
1
Resultater
1
EPA Method 515.3 Laboratory Performance Check Mixture 406 12.5-25 μg/mL in Methyl tert Butyl Ether, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality