CAS RN 185379-40-2
IUPAC Navn:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(pyridin-2-yl)propanoate
Synonymer:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoate
Molekylvægt (g/mol):
387.42
Molekylformel:
C23H19N2O4
InChi Key:
DXIVJCDZOMUITR-NRFANRHFSA-M
SMILES:
[O-]C(=O)[C@H](CC1=CC=CC=N1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
af
1
Resultater
1
Fmoc-l-2-pyridylalanine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.