CAS RN 184377-56-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
p-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(Beta-D-galactopyranosyl)-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
p-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(Beta-D-galactopyranosyl)-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.