CAS RN 1823122-52-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
DOTA-NHS-ester Hexafluorophosphate TFA Salt (Technical Grade), TRC
CAS: 1823122-52-6 Molekylær formel: C20H31N5O10 . C2HF3O2 . HPF6 Synonym: 1,4,7,10-Tetraazacyclododecane-N,N',N' ',N' ' '-tetraacetic acid mono(N-hydroxysuccinimidyl) ester Hexafluorophosphate TFA Salt,1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid mono(N-hydroxysuccinimide ester) Hexafluorophosphate TFA Salt SMIL: OC(=O)CN1CCN(CC(=O)O)CCN(CC(=O)ON2C(=O)CCC2=O)CCN(CC(=O)O)CC1.OC(=O)C(F)(F)F.F[PH](F)(F)(F)(F)F