CAS RN 1821280-29-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
SBI-0640756, MedChemExpress
MedChemExpress SBI-0640756 (SBI-756) is an inhibitor of eIF4G1 and disrupts the eIF4F complex.