CAS RN 1820572-35-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(3aR,4R,7S,7aS)-Octahydro-4,7-methano-1H-isoindole Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3aR,4R,7S,7aS)-Octahydro-4,7-methano-1H-isoindole Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3aR,4R,7S,7aS)-Octahydro-4,7-methano-1H-isoindole Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.