CAS RN 1811539-88-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Tocris Bioscience™ Janelia Fluor 585, NHS ester
Orange dye (Reactive group: NHS ester). Application: confocal microscopy, super resolution microscopy, and two-photon excitation fluorescence microscopy. Suitable for live cell imaging
2
Janelia Fluor 585, SE
Orange dye (Reactive group: NHS ester). Applications: super resolution microscopy, confocal, two-photon excitation and live cell imaging
3
Janelia Fluor 585, SE
Orange dye (Reactive group: NHS ester). Applications: super resolution microscopy, confocal, two-photon excitation and live cell imaging