CAS RN 1808951-93-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ LP 99
Selective BRD7 and BRD9 inhibitor (Kd values are 99 and 909 nM, respectively)