CAS RN 1807350-90-8
IUPAC Navn:
ethyl pyrrolidine-3-carboxylate hydrochloride
Synonymer:
ethyl pyrrolidine-3-carboxylate hydrochloride
Molekylvægt (g/mol):
179.64
Molekylformel:
C7H14ClNO2
InChi Key:
AYYKCALHTPKNOI-UHFFFAOYNA-N
SMILES:
Cl.CCOC(=O)C1CCNC1
1
–
1
af
1
Resultater
1
D-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
CAS: 1807350-90-8 Molekylær formel: C7H14ClNO2 Molekylvægt (g/mol): 179.644 MDL nummer: MFCD18071037 InChI nøgle: AYYKCALHTPKNOI-FYZOBXCZSA-N Synonym: ethyl r-3-pyrrolidinecarboxylate hydrochloride,d-beta-proline ethyl ester hydrochloride,r-ethyl pyrrolidine-3-carboxylate hydrochloride,ethyl r-3-pyrrolidinecarboxylate, hcl,r-ethyl pyrrolidine-3-carboxylate hcl,ethyl r---pyrrolidine-3-carboxylate hydrochloride,ethyl r-pyrrolidine-3-carboxylate hydrochloride,ethyl 3r-pyrrolidine-3-carboxylate hydrochloride,r---pyrrolidine-3-carboxylic acid ethyl ester hydrochloride PubChem CID: 53488453 IUPAC navn: ethyl (3R)-pyrrolidin-3-carboxylat;hydrochlorid SMIL: CCOC(=O)C1CCNC1.Cl