CAS RN 179756-58-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Eptapirone, MedChemExpress
MedChemExpress Eptapirone (F11440) is a potent, selective, high efficacy 5-HT1A receptor agonist with marked anxiolytic and antidepressant potential.