CAS RN 179532-60-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
[3aS-(3aalpha,4beta,6aalpha)]-Hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-Thieno[3,4-d]imidazole-4-pentanoic Acid Phenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[3aS-(3aAlpha,4Beta,6aAlpha)]-Hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-Thieno[3,4-d]imidazole-4-pentanoic Acid Phenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.