CAS RN 17804-35-2
IUPAC Navn:
methyl N-[1-(butylcarbamoyl)-1H-1,3-benzodiazol-2-yl]carbamate
Synonymer:
benomyl benlate arbortrine BBC BNM MBC fundasol NS 02 (fungicide) benomyl (substance) fundazol
Molekylvægt (g/mol):
290.32
Molekylformel:
C14H18N4O3
InChi Key:
RIOXQFHNBCKOKP-UHFFFAOYSA-N
SMILES:
CCCCNC(=O)N1C(NC(=O)OC)=NC2=CC=CC=C12
1
–
4
af
4
Resultater
1
Benomyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benomyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benomyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Benomyl, Fluka™
Analytical standard, reference material