CAS RN 177793-79-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(1S)-N-(3S)-1-Azabicyclo[2.2.2]oct-3-yl-1,2,3,4-tetrahydro-1-naphthalenecarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1S)-N-(3S)-1-Azabicyclo[2.2.2]oct-3-yl-1,2,3,4-tetrahydro-1-naphthalenecarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.