CAS RN 173159-57-4
IUPAC Navn:
2-({[(4,6-dimethoxypyrimidin-2-yl)carbamoyl]amino}sulfonyl)-3-formamido-N,N-dimethylbenzamide
Synonymer:
2-{[(4,6-dimethoxypyrimidin-2-yl)carbamoyl]aminosulfonyl}-3-formamido-N,N-dimethylbenzamide
Molekylvægt (g/mol):
452.44
Molekylformel:
C17H20N6O7S
InChi Key:
HICJAEZEEBOKGV-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=NC(NC(=O)NS(=O)(=O)C2=C(NC=O)C=CC=C2C(=O)N(C)C)=N1
1
–
4
af
4
Resultater
1
Foramsulfuron, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Foramsulfuron, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Foramsulfuron, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.