CAS RN 170809-51-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
MedChemExpress Astressin
MedChemExpress Astressin is a potent corticotropin releasing factor (CRF) antagonist.
2
MedChemExpress Astressin
MedChemExpress Astressin is a potent corticotropin releasing factor (CRF) antagonist.