CAS RN 1704740-52-2
IUPAC Navn:
N-(4-{[3-cyano-1-(2-methylpropyl)-1H-indol-5-yl]oxy}phenyl)pyrrolidine-2-carboxamide
Synonymer:
N-(4-{[3-cyano-1-(2-methylpropyl)indol-5-yl]oxy}phenyl)pyrrolidine-2-carboxamide
Molekylvægt (g/mol):
402.5
Molekylformel:
C24H26N4O2
InChi Key:
OOGHGWKBJXQNEJ-UHFFFAOYNA-N
SMILES:
CC(C)CN1C=C(C#N)C2=CC(OC3=CC=C(NC(=O)C4CCCN4)C=C3)=CC=C12
1
–
2
af
2
Resultater
1
Tocris Bioscience™ AZ PFKFB3 26
CAS: 1704740-52-2 Molekylær formel: C24H26N4O2
2
AZ PFKFB3 26, MedChemExpress
MedChemExpress AZ PFKFB3 26 is a potent and selective inhibitor of the metabolic kinase PFKFB3 with an IC50 of 23 nM. AZ PFKFB3 26 inhibits PFKFB1 and PFKFB2 with IC50s of 2.06 and 0.384 μM, respectively.