CAS RN 1691234-23-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(3Beta,5Alpha,22E)-3-[[(1,1-Dimethylethyl)diphenylsilyl]oxy] Ergosta-7,22-diene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3b,5a,22E)-3-[[(1,1-Dimethylethyl)diphenylsilyl]oxy] Ergosta-7,22-diene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3Beta,5Alpha,22E)-3-[[(1,1-Dimethylethyl)diphenylsilyl]oxy] Ergosta-7,22-diene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.