CAS RN 167684-17-5
IUPAC Navn:
[1,1'-biphenyl]-3,3',4,4'-tetramine hydrate tetrahydrochloride
Synonymer:
3,3'-diaminobenzidine hydrate tetrahydrochloride
Molekylvægt (g/mol):
378.12
Molekylformel:
C12H20Cl4N4O
InChi Key:
DXWSCXIZIHILNP-UHFFFAOYSA-N
SMILES:
O.Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1
1
–
3
af
3
Resultater
1
3,3'-Diaminobenzidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,3'-Diaminobenzidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,3'-Diaminobenzidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.