CAS RN 16605-91-7
IUPAC Navn:
2,3-dichloro-1,1'-biphenyl
Synonymer:
2,3-dichlorobiphenyl
Molekylvægt (g/mol):
223.1
Molekylformel:
C12H8Cl2
InChi Key:
XOMKZKJEJBZBJJ-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(=C1Cl)C1=CC=CC=C1
1
–
2
af
2
Resultater
1
PCB Congeners Mixture 981 100 μg/mL in Isooctane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
PCB Mixture for HJ 350-2007 100 μg/mL in Hexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality